Gene Transcripts

Gene Symbol
Nkain1
Entrez Gene ID
67149
Gene Name
Na+/K+ transporting ATPase interacting 1
Gene Aliases
2610200G18Rik, 2810426C15Rik
Chromosome Location
Chr. 4: 130530131 - 130574036 on Build GRCm38
UniGene ID
Mm.423029
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025998.3 NP_080274.2 7 1055
GenBank mRNA AK011850.1 - 7 986
AK076029.1 - 4 537
BC116391.1 - 7 992 40101865
BC116392.1 - 7 1009
EF058048.2 - 7 1108

View Full Product Details