Gene Transcripts

Gene Symbol
Lonrf2
Entrez Gene ID
381338
Gene Name
LON peptidase N-terminal domain and ring finger 2
Gene Aliases
2900060P06Rik, AI851349
Chromosome Location
Chr. 1: 38793669 - 38836719 on Build GRCm38
UniGene ID
Mm.487137
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029878.1 NP_001025049.1 11 4178
GenBank mRNA AK132156.1 - 1 981
BC096025.1 - 3 2750
BC139127.1 - 11 4229 8862149
BC139130.1 - 11 4229 8862152

View Full Product Details