Gene Transcripts

Gene Symbol
Odf3b
Entrez Gene ID
70113
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
2010001J22Rik
Chromosome Location
Chr. 15: 89377447 - 89379681 on Build GRCm38
UniGene ID
Mm.247273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013022.1 NP_001013040.1 1, 2 185
GenBank mRNA BC087962.1 - 1, 2 185

View Full Product Details