Gene Transcripts

Gene Symbol
Nptxr
Entrez Gene ID
73340
Gene Name
neuronal pentraxin receptor
Gene Aliases
1200009K17Rik, 1700036C17Rik, 5730406O18Rik, AI452369, AI785356, D15Bwg0580e, NPR, Npcd
Chromosome Location
Chr. 15: 79786351 - 79804709 on Build GRCm38
UniGene ID
Mm.38438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030689.4 NP_109614.2 5 4609
GenBank mRNA AK147546.1 - 5 4667
AK147605.1 - 5 4618
BC024758.1 - 1 1895
BC026413.1 - 1 1507
BC048914.1 - 1 1569
BC058962.1 - 5 4608 6826075
BC059870.1 - 1 1905

Gene Symbol
Npcd
Entrez Gene ID
504193
Gene Name
neuronal pentraxin chromo domain
Gene Aliases
Cbx6-Nptxr
Chromosome Location
Chr. 15: 79786351 - 79834333 on Build GRCm38
UniGene ID
Mm.38438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013360.2 NP_001013378.2 8 4242
NM_001013362.2 NP_001013380.2 9 4533
GenBank mRNA BC060093.1 - 1 2046

View Full Product Details