Gene Transcripts

Gene Symbol
Vom1r89
Entrez Gene ID
494276
Gene Name
vomeronasal 1 receptor 89
Gene Aliases
V1rc15
Chromosome Location
Chr. 4: 97566927 - 97567844 on Build Rnor_6.0
UniGene ID
Rn.129651
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008928.1 NP_001008928.1 1 88
GenBank mRNA AY510298.1 - 1 88

View Full Product Details