Gene Transcripts

Gene Symbol
Mtl5
Entrez Gene ID
309142
Gene Name
metallothionein-like 5, testis-specific (tesmin)
Gene Aliases
Chromosome Location
Chr. 1: 218632764 - 218651054 on Build Rnor_6.0
UniGene ID
Rn.105169
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012069.2 NP_001012069.2 7 991
GenBank mRNA BC083920.1 - 7 977

View Full Product Details