Gene Transcripts

Gene Symbol
Odf3b
Entrez Gene ID
70113
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
2010001J22Rik
Chromosome Location
Chr. 15: 89377447 - 89379681 on Build GRCm38
UniGene ID
Mm.247273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013022.1 NP_001013040.1 3 412
GenBank mRNA AK008010.1 - 2 304
BC037508.1 - 1 463
BC087962.1 - 3 412 6748887

View Full Product Details