Gene Transcripts

Gene Symbol
Laptm4b
Entrez Gene ID
315047
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
Chromosome Location
Chr. 7: 72924911 - 72968099 on Build Rnor_6.0
UniGene ID
Rn.8123
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013174.1 NP_001013192.1 6 637
GenBank mRNA BC086442.1 - 6 637
FQ234072.1 - 2 81

View Full Product Details