Gene Transcripts

Gene Symbol
Pars2
Entrez Gene ID
313429
Gene Name
prolyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
RGD1305345
Chromosome Location
Chr. 5: 126254125 - 126259204 on Build Rnor_6.0
UniGene ID
Rn.205937
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014064.1 NP_001014086.1 2 2401
GenBank mRNA BC088403.1 - 2 2401

View Full Product Details