Gene Transcripts

Gene Symbol
Nostrin
Entrez Gene ID
311111
Gene Name
nitric oxide synthase trafficking
Gene Aliases
Chromosome Location
Chr. 3: 55369282 - 55433840 on Build Rnor_6.0
UniGene ID
Rn.39383
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024260.1 NP_001019431.1 11 1001
GenBank mRNA BC088446.1 - 11 1001

View Full Product Details