Gene Transcripts

Gene Symbol
Eif2d
Entrez Gene ID
498225
Gene Name
eukaryotic translation initiation factor 2D
Gene Aliases
Lgtn
Chromosome Location
Chr. 13: 47935139 - 47958459 on Build Rnor_6.0
UniGene ID
Rn.162387
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017489.1 NP_001017489.1 7 1014
XM_006249790.2 XP_006249852.1 7 1038
GenBank mRNA BC087701.1 - 7 1014

View Full Product Details