Gene Transcripts

Gene Symbol
Rbmxl2
Entrez Gene ID
76572
Gene Name
RNA binding motif protein, X-linked-like 2
Gene Aliases
1700012H05Rik, Hnrnpg-t
Chromosome Location
Chr. 7: 107209445 - 107210916 on Build GRCm38
UniGene ID
Mm.128134
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029660.2 NP_083936.1 1 1302
GenBank mRNA AK005907.1 - 1 1303

View Full Product Details