Gene Transcripts

Gene Symbol
RIIAD1
Entrez Gene ID
284485
Gene Name
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Gene Aliases
C1orf230, NCRNA00166
Chromosome Location
Chr. 1: 151721537 - 151729606 on Build GRCh38
UniGene ID
Hs.297967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144956.1 NP_001138428.1 4 226
GenBank mRNA AA393535.1 - 4 216

View Full Product Details