Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042575.2 | NP_001036040.2 | 5 | 562 | |
NR_026734.1 | 6 | 675 | |||
XM_011512754.1 | XP_011511056.1 | 6 | 1007 | ||
XM_011512758.1 | XP_011511060.1 | 5 | 428 | ||
GenBank mRNA | AK131211.1 | - | 6 | 675 | |
BC117322.1 | - | 5 | 474 | 40125873 |