Gene Transcripts

Gene Symbol
PLSCR5
Entrez Gene ID
389158
Gene Name
phospholipid scramblase family member 5
Gene Aliases
Chromosome Location
Chr. 3: 146576555 - 146606216 on Build GRCh38
UniGene ID
Hs.254692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085420.1 NP_001078889.1 2, 3 1197
XM_017006373.1 XP_016861862.1 2, 3 408
GenBank mRNA AY436642.1 - 2, 3 1197

View Full Product Details