Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001105663.2 | NP_001099133.1 | 2 | 159 | |
NM_001243657.1 | NP_001230586.1 | 2 | 159 | ||
NM_001243660.1 | NP_001230589.1 | 2 | 159 | ||
NM_001243661.1 | NP_001230590.1 | 2 | 159 | ||
XM_011523032.2 | XP_011521334.1 | 2 | 163 | ||
XM_011523033.2 | XP_011521335.1 | 2 | 163 | ||
XM_017023170.1 | XP_016878659.1 | 2 | 163 | ||
GenBank mRNA | AA227330.1 | - | 2 | 111 | |
AK296963.1 | - | 2 | 132 | ||
BU579534.1 | - | 2 | 105 | ||
BX387030.2 | - | 2 | 159 | ||
BX494638.1 | - | 2 | 138 | ||
CN297379.1 | - | 2 | 110 | ||
EU981826.1 | - | 2 | 68 |