Gene Transcripts

Gene Symbol
TRIM71
Entrez Gene ID
131405
Gene Name
tripartite motif containing 71
Gene Aliases
LIN-41, LIN41
Chromosome Location
Chr. 3: 32818018 - 32897826 on Build GRCh38
UniGene ID
Hs.113170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039111.2 NP_001034200.1 4 1301
GenBank mRNA DQ232881.1 - 4 1301

View Full Product Details