Gene Transcripts

Gene Symbol
SIRPB2
Entrez Gene ID
284759
Gene Name
signal regulatory protein beta 2
Gene Aliases
PTPN1L, PTPNS1L3, dJ776F14.2
Chromosome Location
Chr. 20: 1474591 - 1491762 on Build GRCh38
UniGene ID
Hs.721685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122962.1 NP_001116434.1 5 2227
NM_001134836.1 NP_001128308.1 5 1933
GenBank mRNA AK095499.1 - 5 2036
BG539998.1 - 1 389

View Full Product Details