Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018924.4 | NP_061747.2 | 1 | 418 | |
NM_032097.2 | NP_115268.2 | 1 | 418 | ||
GenBank mRNA | AF152332.1 | - | 1 | 287 | |
AF152519.1 | - | 1 | 287 | ||
BC150172.1 | - | 1 | 418 | 40147143 |