Gene Transcripts

Gene Symbol
METTL21C
Entrez Gene ID
196541
Gene Name
methyltransferase like 21C
Gene Aliases
C13orf39
Chromosome Location
Chr. 13: 102685747 - 102704311 on Build GRCh38
UniGene ID
Hs.508623
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010977.2 NP_001010977.1 2 225
XM_017020407.1 XP_016875896.1 3 1119
XM_017020408.1 XP_016875897.1 3 1672
GenBank mRNA BC132748.1 - 2 225
BC132750.1 - 2 225 40146772

View Full Product Details