Gene Transcripts

Gene Symbol
LRRC24
Entrez Gene ID
441381
Gene Name
leucine rich repeat containing 24
Gene Aliases
LRRC14OS
Chromosome Location
Chr. 8: 144522377 - 144527032 on Build GRCh38
UniGene ID
Hs.740859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024678.3 NP_001019849.2 4 587
GenBank mRNA AB178281.1 - 3 518
BC111067.1 - 4 579 5767723

View Full Product Details