Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014976.1 | NP_055791.1 | 29 | 4329 | |
XM_005269647.3 | XP_005269704.1 | 29 | 4369 | ||
XM_011539538.2 | XP_011537840.1 | 29 | 4317 | ||
XM_011539539.2 | XP_011537841.1 | 29 | 4317 | ||
XM_011539540.1 | XP_011537842.1 | 28 | 4183 | ||
GenBank mRNA | AK294971.1 | - | 3968 | ||
BC049838.1 | - | 29 | 4330 | ||
BC064486.1 | - | 29 | 4322 | ||
BC080560.1 | - | 29 | 4322 | ||
BC111040.1 | - | 29 | 4284 | ||
D80007.1 | - | 29 | 4282 |