Gene Transcripts

Gene Symbol
Ly49s6
Entrez Gene ID
494196
Gene Name
Ly49 stimulatory receptor 6
Gene Aliases
Chromosome Location
Chr. 4: 164429048 - 164453171 on Build Rnor_6.0
UniGene ID
Rn.126999
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009488.1 NP_001009488.1 2 193
GenBank mRNA AY649837.1 - 2 193
BC088257.1 - 2 244 7375677

View Full Product Details