Gene Transcripts
- Gene Symbol
- Ly49i5
- Entrez Gene ID
- 494211
- Gene Name
- Ly49 inhibitory receptor 5
- Gene Aliases
- Chromosome Location
- Chr. 4: 164345763 - 164357619 on Build Rnor_6.0
- UniGene ID
- Rn.154805
- Species
- Rattus norvegicus
- Gene Symbol
- Ly49s5
- Entrez Gene ID
- 503662
- Gene Name
- Ly49 stimulatory receptor 5
- Gene Aliases
- Chromosome Location
- Chr. 4: 164284910 - 164306500 on Build Rnor_6.0
- UniGene ID
- Rn.154805
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY846265.1 |
- |
3 |
396 |
|
View Full Product Details