Gene Transcripts

Gene Symbol
Msl3l2
Entrez Gene ID
309790
Gene Name
male-specific lethal 3-like 2 (Drosophila)
Gene Aliases
RGD1308699
Chromosome Location
Chr. 20: 37580068 - 37590015 on Build Rnor_6.0
UniGene ID
Rn.230094
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014032.1 NP_001014054.1 1 1127
XM_006256504.2 XP_006256566.1 3 2709
GenBank mRNA BC079056.1 - 1 1127

View Full Product Details