Gene Transcripts

Gene Symbol
Ndufb4
Entrez Gene ID
288088
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4
Gene Aliases
Chromosome Location
Chr. 11: 65960277 - 65966899 on Build Rnor_6.0
UniGene ID
Rn.98986
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037338.1 NP_001032415.1 3 419
GenBank mRNA CB732095.1 - 3 419
DQ255901.1 - 3 272
FQ217067.1 - 3 426

View Full Product Details