Gene Transcripts

Gene Symbol
Nkapl
Entrez Gene ID
291165
Gene Name
NFKB activating protein-like
Gene Aliases
RGD1311667
Chromosome Location
Chr. 17: 45175254 - 45176771 on Build Rnor_6.0
UniGene ID
Rn.104854
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029913.1 NP_001025084.1 1 569
GenBank mRNA BC097473.1 - 1 569

View Full Product Details