Gene Transcripts

Gene Symbol
Memo1
Entrez Gene ID
298787
Gene Name
mediator of cell motility 1
Gene Aliases
RGD1309929
Chromosome Location
Chr. 6: 22362483 - 22409583 on Build Rnor_6.0
UniGene ID
Rn.22312
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029917.1 NP_001025088.1 8 808
GenBank mRNA BC098061.1 - 8 808
FQ215386.1 - 6 560

View Full Product Details