Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024622.2 | NP_001019793.1 | 2 | 138 | |
GenBank mRNA | AK145195.1 | - | 2 | 97 | |
AK150595.1 | - | 2 | 99 | ||
AK152279.1 | - | 2 | 101 | ||
AK159201.1 | - | 2 | 109 | ||
AK166071.1 | - | 2 | 122 | ||
AK172325.1 | - | 1 | 143 | ||
AK189299.1 | - | 1 | 227 | ||
BC061235.1 | - | 2 | 102 | 6593866 | |
BC132588.1 | - | 2 | 160 | 40130865 | |
BC132590.1 | - | 2 | 160 | 40130867 | |
BY287179.1 | - | 2 | 138 |