Gene Transcripts
- Gene Symbol
- 4933402N22Rik
- Entrez Gene ID
- 545732
- Gene Name
- RIKEN cDNA 4933402N22 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 11918043 - 11922804 on Build GRCm38
- UniGene ID
- Mm.389066
- Species
- Mus musculus
- Gene Symbol
- Gm5861
- Entrez Gene ID
- 545728
- Gene Name
- predicted gene 5861
- Gene Aliases
- EG545728
- Chromosome Location
- Chr. 5: 11182919 - 11187810 on Build GRCm38
- UniGene ID
- Mm.109461
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC030042.1 |
- |
|
462 |
|
- Gene Symbol
- Gm6455
- Entrez Gene ID
- 623849
- Gene Name
- predicted gene 6455
- Gene Aliases
- EG623849
- Chromosome Location
- Chr. 5: 10865029 - 10870808 on Build GRCm38
- UniGene ID
- Mm.335888
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC048648.1 |
- |
4 |
434 |
|
- Gene Symbol
- Gm6460
- Entrez Gene ID
- 623898
- Gene Name
- predicted gene 6460
- Gene Aliases
- EG623898
- Chromosome Location
- Chr. 5: 11594956 - 11599480 on Build GRCm38
- UniGene ID
- Mm.425228
- Species
- Mus musculus
- Gene Symbol
- Speer1
- Entrez Gene ID
- 70896
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 1
- Gene Aliases
- 4921519H07Rik, SPEER-1, Speer1-ps, Speer1-ps1
- Chromosome Location
- Chr. 5: 11340408 - 11346273 on Build GRCm38
- UniGene ID
- Mm.425218
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY151400.1 |
- |
|
328 |
|
View Full Product Details