Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001015002.1 | NP_001015002.1 | 9 | 1006 | |
NM_001031803.1 | NP_001026973.1 | 9 | 1006 | ||
NM_004524.2 | NP_004515.2 | 9 | 1006 | ||
XM_011524801.1 | XP_011523103.1 | 9 | 989 | ||
XM_011524802.1 | XP_011523104.1 | 9 | 977 | ||
XM_011524807.2 | XP_011523109.1 | 9 | 1094 | ||
XM_017024625.1 | XP_016880114.1 | 9 | 977 | ||
XM_017024626.1 | XP_016880115.1 | 9 | 977 | ||
XM_017024627.1 | XP_016880116.1 | 9 | 977 | ||
XM_017024628.1 | XP_016880117.1 | 9 | 977 | ||
XM_017024629.1 | XP_016880118.1 | 9 | 977 | ||
XM_017024630.1 | XP_016880119.1 | 9 | 977 | ||
XM_017024631.1 | XP_016880120.1 | 9 | 977 | ||
GenBank mRNA | AK126687.1 | - | 6 | 1029 | |
AK315126.1 | - | 9 | 956 | ||
BC006503.2 | - | 10 | 1064 | ||
BC010879.1 | - | 9 | 1007 | ||
BC064994.1 | - | 9 | 979 | 5575350 | |
X87342.1 | - | 9 | 982 |