Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022042.3 | NP_071325.2 | 2 | 549 | |
NM_134425.2 | NP_602297.1 | 2 | 549 | ||
NM_213613.3 | NP_998778.1 | 3 | 764 | ||
GenBank mRNA | AF297659.2 | - | 2 | 504 | |
AK292747.1 | - | 2 | 549 | ||
AY124771.1 | - | 3 | 760 | ||
BC015517.2 | - | 2 | 485 |