Gene Transcripts

Gene Symbol
OR2H2
Entrez Gene ID
7932
Gene Name
olfactory receptor family 2 subfamily H member 2
Gene Aliases
FAT11, OLFR2, OLFR42B, OR2H3, dJ271M21.2, hs6M1-12
Chromosome Location
Chr. 6: 29586524 - 29591022 on Build GRCh38
UniGene ID
Hs.529493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007160.3 NP_009091.3 1 1018
GenBank mRNA BC101683.1 - 1 1018
BC112176.1 - 1 1030 8327644

View Full Product Details