Gene Transcripts

Gene Symbol
MS4A7
Entrez Gene ID
58475
Gene Name
membrane spanning 4-domains A7
Gene Aliases
4SPAN2, CD20L4, CFFM4, MS4A8
Chromosome Location
Chr. 11: 60378485 - 60395954 on Build GRCh38
UniGene ID
Hs.530735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021201.4 NP_067024.1 7 943
NM_206938.1 NP_996821.1 6 808
NM_206939.1 NP_996822.1 7 895
NM_206940.1 NP_996823.1 6 760
GenBank mRNA AB026043.1 - 7 883
AF201951.1 - 7 854
AF237916.1 - 7 867
AF309653.1 - 7 896
AF350501.1 - 7 854
AK075106.1 - 7 947
AK297027.1 - 6 744
BC020673.1 - 7 867 4720044

View Full Product Details