Gene Transcripts

Gene Symbol
NOL6
Entrez Gene ID
65083
Gene Name
nucleolar protein 6
Gene Aliases
NRAP, UTP22, bA311H10.1
Chromosome Location
Chr. 9: 33460811 - 33473943 on Build GRCh38
UniGene ID
Hs.493709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022917.4 NP_075068.2 2 219
NM_139235.3 NP_631981.2 2 219
XM_017015044.1 XP_016870533.1 2 155
GenBank mRNA AF361079.1 - 2 177
AF361080.1 - 2 177
AF361081.1 - 2 177
AK026258.1 - 2 178
AK293969.1 - 2 178
BC020450.1 - 2 164
BC030139.2 - 2 132 4423152
CB267269.1 - 2 219

Gene Symbol
SUGT1P1
Entrez Gene ID
441394
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone pseudogene 1
Gene Aliases
SGT1P, SUGT1P, bA255A11.1
Chromosome Location
Chr. 9: 33500756 - 33511166 on Build GRCh38
UniGene ID
Hs.521441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA DA804368.1 - 7 534

View Full Product Details