Gene Transcripts

Gene Symbol
PARP8
Entrez Gene ID
79668
Gene Name
poly(ADP-ribose) polymerase family member 8
Gene Aliases
ARTD16, pART16
Chromosome Location
Chr. 5: 50665899 - 50846522 on Build GRCh38
UniGene ID
Hs.369581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178055.1 NP_001171526.1 6, 7 535
NM_001178056.1 NP_001171527.1 5, 6 511
NM_024615.3 NP_078891.2 5, 6 511
XM_005248596.4 XP_005248653.1 6, 7 891
XM_011543631.2 XP_011541933.1 6, 7 424
XM_011543632.2 XP_011541934.2 6, 7 471
XM_011543633.2 XP_011541935.2 6, 7 471
XM_011543634.2 XP_011541936.1 7, 8 1020
XM_011543635.2 XP_011541937.1 7, 8 541
XM_011543636.2 XP_011541938.1 6, 7 405
XM_011543640.2 XP_011541942.1 5, 6 447
XM_011543641.2 XP_011541943.1 6, 7 424
XM_017009850.1 XP_016865339.1 5, 6 971
GenBank mRNA AK289369.1 - 5, 6 494
AK314466.1 - 6, 7 535
AL834477.1 - 5, 6 470
BC075801.1 - 5, 6 470 5226828
BC105789.1 - 6, 7 539
DC417471.1 - 5, 6 511
DC429180.1 - 5, 6 483

View Full Product Details