Gene Transcripts

Gene Symbol
PARP8
Entrez Gene ID
79668
Gene Name
poly(ADP-ribose) polymerase family member 8
Gene Aliases
ARTD16, pART16
Chromosome Location
Chr. 5: 50665899 - 50846522 on Build GRCh38
UniGene ID
Hs.369581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178055.1 NP_001171526.1 7 562
NM_001178056.1 NP_001171527.1 6 538
NM_024615.3 NP_078891.2 6 538
XM_005248596.4 XP_005248653.1 7 918
XM_011543631.2 XP_011541933.1 7 451
XM_011543632.2 XP_011541934.2 7 498
XM_011543633.2 XP_011541935.2 7 498
XM_011543634.2 XP_011541936.1 8 1047
XM_011543635.2 XP_011541937.1 8 568
XM_011543636.2 XP_011541938.1 7 432
XM_011543640.2 XP_011541942.1 6 474
XM_011543641.2 XP_011541943.1 7 451
XM_011543642.2 XP_011541944.1 5 438
XM_011543643.2 XP_011541945.1 6 879
XM_017009850.1 XP_016865339.1 6 998
XM_017009851.1 XP_016865340.1 5 438
GenBank mRNA AK289369.1 - 6 521
AK298680.1 - 5 401
AK314466.1 - 7 562
AL834477.1 - 6 497
BC037386.1 - 4 396 4830820
BC075801.1 - 6 497 5226828
BC105789.1 - 7 566
DC429180.1 - 6 510

View Full Product Details