Gene Transcripts

Gene Symbol
OGFOD2
Entrez Gene ID
79676
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 2
Gene Aliases
Chromosome Location
Chr. 12: 122974703 - 122980041 on Build GRCh38
UniGene ID
Hs.524817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304833.1 NP_001291762.1 7 1470
NM_001304834.1 NP_001291763.1 8 1901
NM_001304835.1 NP_001291764.1 7 2021
NM_001304836.1 NP_001291765.1 7 1790
NM_001304837.1 NP_001291766.1 6 1651
NM_001304838.1 NP_001291767.1 7 1708
NM_024623.2 NP_078899.1 8 1857
GenBank mRNA AK023553.1 - 8 1753
AK094820.1 - 7 1453
AK095481.1 - 6 1559
BC098119.1 - 8 1633 40024912
BC098293.1 - 8 1633 40024914
BC101940.1 - 7 1440 40024915
BC105637.1 - 7 1440 40024913
BX640734.1 - 8 1501

View Full Product Details