Gene Transcripts

Gene Symbol
MAP6D1
Entrez Gene ID
79929
Gene Name
MAP6 domain containing 1
Gene Aliases
MAPO6D1, SL21
Chromosome Location
Chr. 3: 183815876 - 183825605 on Build GRCh38
UniGene ID
Hs.478465
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024871.2 NP_079147.1 2 471
GenBank mRNA AK022810.1 - 2 443
BC006434.1 - 2 436 3937613

View Full Product Details