Gene Transcripts

Gene Symbol
LY6G5C
Entrez Gene ID
80741
Gene Name
lymphocyte antigen 6 complex, locus G5C
Gene Aliases
C6orf20, G5C, LY6G5CA, LY6G5CB, NG33
Chromosome Location
Chr. 6: 31676684 - 31680373 on Build GRCh38
UniGene ID
Hs.25738
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025262.3 NP_079538.3 3 444
GenBank mRNA AA179679.1 - 1 86

View Full Product Details