Gene Transcripts
- Gene Symbol
- MXRA8
- Entrez Gene ID
- 54587
- Gene Name
- matrix remodeling associated 8
- Gene Aliases
- Chromosome Location
- Chr. 1: 1352689 - 1363541 on Build GRCh38
- UniGene ID
- Hs.515687
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282582.1 |
NP_001269511.1
|
10, 11 |
1379 |
|
NM_001282583.1 |
NP_001269512.1
|
9, 10 |
1798 |
|
NM_001282584.1 |
NP_001269513.1
|
8, 9 |
1298 |
|
NM_032348.3 |
NP_115724.1
|
9, 10 |
1601 |
|
GenBank mRNA |
AB052096.1 |
- |
10, 11 |
1379 |
|
AK095966.1 |
- |
9, 10 |
1798 |
|
AK293442.1 |
- |
8, 9 |
1067 |
|
AK304615.1 |
- |
8, 9 |
1393 |
|
AK315510.1 |
- |
9, 10 |
1368 |
|
AL050202.1 |
- |
3, 4 |
237 |
|
AY358391.1 |
- |
8, 9 |
1044 |
|
BC006213.2 |
- |
9, 10 |
1342 |
3343261 |
BC017312.2 |
- |
9, 10 |
1342 |
5090754 |
View Full Product Details