Gene Transcripts

Gene Symbol
TMEM60
Entrez Gene ID
85025
Gene Name
transmembrane protein 60
Gene Aliases
C7orf35, DC32
Chromosome Location
Chr. 7: 77793728 - 77798430 on Build GRCh38
UniGene ID
Hs.19025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032936.3 NP_116325.1 2 732
GenBank mRNA AF260336.1 - 2 733
AK095664.1 - 3887
BC065930.1 - 2 656 6146561

View Full Product Details