Gene Transcripts

Gene Symbol
SEC16B
Entrez Gene ID
89866
Gene Name
SEC16 homolog B, endoplasmic reticulum export factor
Gene Aliases
LZTR2, PGPR-p117, RGPR, SEC16S
Chromosome Location
Chr. 1: 177929107 - 177969915 on Build GRCh38
UniGene ID
Hs.149540
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033127.2 NP_149118.2 7, 8 1034
GenBank mRNA AB063357.1 - 7, 8 1034
BC009106.1 - 7, 8 1029 3448742
EF125213.1 - 6, 7 944

View Full Product Details