Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146728.1 | NP_001140200.1 | 3 | 531 | |
NM_001146729.1 | NP_001140201.1 | 3 | 501 | ||
NM_054108.3 | NP_473449.1 | 3 | 531 | ||
XM_011544751.2 | XP_011543053.1 | 3 | 436 | ||
XM_011544753.1 | XP_011543055.1 | 3 | 264 | ||
XM_017017172.1 | XP_016872661.1 | 3 | 252 | ||
GenBank mRNA | AB298804.1 | - | 3 | 357 | |
AJ416558.1 | - | 3 | 518 | ||
AK302312.1 | - | 3 | 492 | ||
BC034222.1 | - | 3 | 460 | 5268703 |