Gene Transcripts

Gene Symbol
NEU4
Entrez Gene ID
129807
Gene Name
neuraminidase 4
Gene Aliases
Chromosome Location
Chr. 2: 241809065 - 241817413 on Build GRCh38
UniGene ID
Hs.551747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167599.2 NP_001161071.1 4 2146
NM_001167600.2 NP_001161072.1 4 1823
NM_001167601.2 NP_001161073.1 4 1826
NM_001167602.2 NP_001161074.1 5 2324
NM_080741.3 NP_542779.2 4 2143
GenBank mRNA AK025617.1 - 5 2076
AK091038.1 - 5 2321
AK096992.1 - 2 2936
AK289421.1 - 4 1700
AK290076.1 - 4 1683
AK298403.1 - 5 1655
AK300969.1 - 4 1696
AY203952.1 - 4 1678
BC012899.1 - 4 2097 4156395
BC105974.1 - 4 1659 4944243

View Full Product Details