Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138697.3 | NP_619642.2 | 2 | 356 | |
NM_177540.2 | NP_803884.1 | 2 | 356 | ||
XM_011542203.2 | XP_011540505.1 | 2 | 722 | ||
XM_011542206.2 | XP_011540508.1 | 1 | 908 | ||
XM_017002402.1 | XP_016857891.1 | 1 | 908 | ||
XM_017002403.1 | XP_016857892.1 | 1 | 908 | ||
GenBank mRNA | AF387617.1 | - | 1 | 37 | |
AF387618.1 | - | 1 | 69 | ||
AF387619.1 | - | 1 | 37 | ||
AF387620.1 | - | 1 | 37 | ||
AK292014.1 | - | 2 | 325 | ||
AK293499.1 | - | 2 | 225 | ||
AK315805.1 | - | 2 | 325 | ||
BC136515.1 | - | 2 | 356 | 9020505 | |
BC136516.1 | - | 2 | 355 | 9020506 |