Gene Transcripts
- Gene Symbol
- PACRG
- Entrez Gene ID
- 135138
- Gene Name
- PARK2 coregulated
- Gene Aliases
- GLUP, HAK005771, PARK2CRG
- Chromosome Location
- Chr. 6: 162726870 - 163315492 on Build GRCh38
- UniGene ID
- Hs.25791
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080378.1 |
NP_001073847.1
|
3, 4 |
523 |
|
NM_001080379.1 |
NP_001073848.1
|
2, 3 |
565 |
|
NM_152410.2 |
NP_689623.2
|
3, 4 |
523 |
|
XM_005266825.4 |
XP_005266882.1
|
2, 3 |
648 |
|
XM_005266826.4 |
XP_005266883.1
|
2, 3 |
648 |
|
XM_006715344.3 |
XP_006715407.1
|
2, 3 |
648 |
|
XM_006715345.3 |
XP_006715408.1
|
2, 3 |
648 |
|
XM_011535461.2 |
XP_011533763.1
|
2, 3 |
648 |
|
XM_011535462.2 |
XP_011533764.1
|
2, 3 |
648 |
|
XM_011535466.1 |
XP_011533768.1
|
2, 3 |
648 |
|
XM_011535468.2 |
XP_011533770.1
|
2, 3 |
648 |
|
XM_011535469.2 |
XP_011533771.1
|
2, 3 |
648 |
|
XM_017010275.1 |
XP_016865764.1
|
3, 4 |
523 |
|
XM_017010276.1 |
XP_016865765.1
|
3, 4 |
514 |
|
XM_017010277.1 |
XP_016865766.1
|
3, 4 |
798 |
|
XM_017010278.1 |
XP_016865767.1
|
3, 4 |
448 |
|
XM_017010279.1 |
XP_016865768.1
|
3, 4 |
475 |
|
XM_017010280.1 |
XP_016865769.1
|
2, 3 |
648 |
|
XM_017010281.1 |
XP_016865770.1
|
1, 2 |
451 |
|
XM_017010282.1 |
XP_016865771.1
|
2, 3 |
367 |
|
GenBank mRNA |
AF546872.1 |
- |
2, 3 |
565 |
|
AK057286.1 |
- |
3, 4 |
523 |
|
BC030642.2 |
- |
2, 3 |
535 |
4823539 |
BC044227.1 |
- |
3, 4 |
510 |
5187302 |
View Full Product Details