Gene Transcripts

Gene Symbol
TTK
Entrez Gene ID
7272
Gene Name
TTK protein kinase
Gene Aliases
CT96, ESK, MPH1, MPS1, MPS1L1, PYT
Chromosome Location
Chr. 6: 80004147 - 80043577 on Build GRCh38
UniGene ID
Hs.169840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166691.1 NP_001160163.1 5 725
NM_003318.4 NP_003309.2 5 713
XM_011536099.2 XP_011534401.1 5 687
XM_011536100.2 XP_011534402.1 5 713
XM_017011242.1 XP_016866731.1 6 1242
GenBank mRNA AK095599.1 - 5 1185
AK292460.1 - 5 688
AK315696.1 - 5 630
AX775733.1 - 5 685
BC000633.2 - 5 676 3343925
BC032858.2 - 5 678 5270012

View Full Product Details