Gene Transcripts

Gene Symbol
SDK1
Entrez Gene ID
221935
Gene Name
sidekick cell adhesion molecule 1
Gene Aliases
Chromosome Location
Chr. 7: 3301448 - 4269000 on Build GRCh38
UniGene ID
Hs.653013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079653.1 NP_001073121.1 18 2643
NM_152744.3 NP_689957.3 44 6498
XM_011515188.1 XP_011513490.1 38 5339
XM_011515189.1 XP_011513491.1 38 5336
XM_017011837.1 XP_016867326.1 38 5339
GenBank mRNA AK055987.1 - 5 569
AK074077.1 - 11 1438
AK074083.1 - 29 4134
AL833354.1 - 18 2640
AY310398.1 - 44 6498

View Full Product Details