Gene Transcripts
- Gene Symbol
- MFSD8
- Entrez Gene ID
- 256471
- Gene Name
- major facilitator superfamily domain containing 8
- Gene Aliases
- CCMD, CLN7
- Chromosome Location
- Chr. 4: 127917805 - 127965984 on Build GRCh38
- UniGene ID
- Hs.480701
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152778.2 |
NP_689991.1
|
5, 6 |
610 |
|
XM_005262893.1 |
XP_005262950.1
|
4, 5 |
524 |
|
XM_005262896.1 |
XP_005262953.1
|
4, 5 |
524 |
|
XM_005262897.2 |
XP_005262954.1
|
4, 5 |
524 |
|
XM_005262898.2 |
XP_005262955.1
|
4, 5 |
524 |
|
XM_017007989.1 |
XP_016863478.1
|
4, 5 |
524 |
|
GenBank mRNA |
AK074564.1 |
- |
4, 5 |
459 |
|
AK294527.1 |
- |
4, 5 |
433 |
|
AK294957.1 |
- |
4, 5 |
452 |
|
AK315596.1 |
- |
4, 5 |
456 |
|
BC029503.1 |
- |
5, 6 |
575 |
5272664 |
BI553701.1 |
- |
4, 5 |
524 |
|
View Full Product Details